missing translation for 'onlineSavingsMsg'
Learn More

4-Acryloylmorpholine (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. A0841500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0841500ML 500 mL
A084125ML 25 mL
A0841100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0841500ML Supplier TCI America Supplier No. A0841500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5117-12-4
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00047413
InChI Key XLPJNCYCZORXHG-UHFFFAOYSA-N
Synonym 4-acryloylmorpholine, 1-morpholinoprop-2-en-1-one, n-acrylylmorpholine, n-acryloylmorpholine, morpholine, 4-1-oxo-2-propenyl, poly acryloylmorpholine, acryloylmorpholine, acryloyl morpholine, 1-morpholin-4-yl prop-2-en-1-one, unii-k0y58p61ja
PubChem CID 98723
IUPAC Name 1-morpholin-4-ylprop-2-en-1-one
SMILES C=CC(=O)N1CCOCC1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 500 mL
Formula Weight 141.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Acryloylmorpholine (stabilized with MEHQ)
EINECSNumber (5)-0865
TSCA Yes