missing translation for 'onlineSavingsMsg'
Learn More

4-Acryloylmorpholine, 98+%, stab. with 4-methoxyphenol

Catalog No. p-7046055
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1294706 5 g
AAL1294714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1294706 Supplier Thermo Scientific Chemicals Supplier No. L1294706
Only null left

CAS: 5117-12-4 | C7H11NO2 | 141.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5117-12-4
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00047413
InChI Key XLPJNCYCZORXHG-UHFFFAOYSA-N
Synonym 4-acryloylmorpholine, 1-morpholinoprop-2-en-1-one, n-acrylylmorpholine, n-acryloylmorpholine, morpholine, 4-1-oxo-2-propenyl, poly acryloylmorpholine, acryloylmorpholine, acryloyl morpholine, 1-morpholin-4-yl prop-2-en-1-one, unii-k0y58p61ja
PubChem CID 98723
IUPAC Name 1-morpholin-4-ylprop-2-en-1-one
SMILES C=CC(=O)N1CCOCC1

Specifications

Melting Point -8°C
Density 1.12
Boiling Point 158°C (50 mmHg)
Flash Point 127°C (260°F)
Refractive Index 1.509
Quantity 5 g
Beilstein 119302
Sensitivity Light sensitive
Formula Weight 141.17
Percent Purity ≥98%
Chemical Name or Material 4-Acryloylmorpholine, stabilized with 4-methoxyphenol
Show More Show Less
Hazard Category H302-H317-H318-H373
Hazard Statement GHS H Statement
H301-H318-H373-H317
Toxic if swallowed.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P333+P313-P363-P501c
EINECSNumber 418-140-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only