missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylphenylboronic acid pinacol ester, 97%

Catalog No. AC444430050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444430050 5 g
AC444430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444430050 Supplier Thermo Scientific Chemicals Supplier No. 444430050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 171364-81-1 | C14H19BO3 | 246.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 171364-81-1
Molecular Formula C14H19BO3
Molecular Weight (g/mol) 246.11
MDL Number MFCD05863922
InChI Key BATKIZWNRQGSKE-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetylphenylboronic acid pinacol ester, 4-acetylphenylboronic acid, pinacol ester, 2-4-acetylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethan-1-one, amtb682, 4-acetylphenyl boronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2yl phenyl-ethanone
PubChem CID 2760596
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 59°C to 62°C
Color White
Boiling Point 80°C (0.1 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Formula Weight 246.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetylphenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only