missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$37.29 - $37.29
Specifications
| CAS | 149104-90-5 |
|---|---|
| Melting Point | 239°C |
| Color | White-Yellow |
| Molecular Formula | C8H9BO3 |
| InChI Key | OBQRODBYVNIZJU-UHFFFAOYSA-N |
Specifications
| 149104-90-5 | |
| White-Yellow | |
| OBQRODBYVNIZJU-UHFFFAOYSA-N | |
| 3702122 | |
| Crystalline Powder |
| 239°C | |
| C8H9BO3 | |
| (4-acetylphenyl)boronic acid | |
| 163.97 |