missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. A19071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19071G Supplier TCI America Supplier No. A19071G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Acetylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 149104-90-5
Melting Point 239°C
Color White-Yellow
Molecular Formula C8H9BO3
MDL Number MFCD01074667
Quantity 1 g
Synonym 4-acetylphenyl boronic acid, p-acetylphenylboronic acid, 4-acetylbenzeneboronic acid, 1-4-dihydroxyboranyl phenyl ethan-1-one, boronic acid, 4-acetylphenyl, 4acetylphenylboronic acid, 4-actylphenylboronic acid, 4acetylbenzeneboronic acid, p-acetylbenzeneboronic acid
InChI Key OBQRODBYVNIZJU-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)B(O)O
IUPAC Name (4-acetylphenyl)boronic acid
Molecular Weight (g/mol) 163.97
PubChem CID 3702122
Formula Weight 163.97
Physical Form Crystalline Powder
Show More Show Less