missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyldiphenyl Sulfide 98.0+%, TCI America™

Catalog No. p-7117857
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23555G 5 g
A235525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23555G Supplier TCI America Supplier No. A23555G
Only null left

Chemical Identifiers

CAS 10169-55-8
Molecular Formula C14H12OS
Molecular Weight (g/mol) 228.31
MDL Number MFCD00026227
InChI Key XUDYHODVSUXRPW-UHFFFAOYSA-N
Synonym 4-acetyldiphenyl sulfide, 1-4-phenylthio phenyl ethanone, 4-acetyl diphenyl sulfide, 4'-phenylthio acetophenone, 1-4-phenylsulfanyl phenyl ethanone, 1-4-phenylsulfanylphenyl ethanone, 1-4-phenylthio phenyl ethan-1-one, ethanone, 1-4-phenylthio phenyl, 1-acetyl-4-phenylthiobenzene
PubChem CID 66287
IUPAC Name 1-[4-(phenylsulfanyl)phenyl]ethan-1-one
SMILES CC(=O)C1=CC=C(SC2=CC=CC=C2)C=C1

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 184°C
Quantity 5 g
Formula Weight 228.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetyldiphenyl Sulfide