missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetyldiphenyl Sulfide 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 67°C |
| Color | White-Yellow |
| Boiling Point | 184°C |
| Quantity | 5 g |
| Formula Weight | 228.31 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Acetyldiphenyl Sulfide |
Sécurité et manipulation
| TSCA | No |