missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbiphenyl 98.0+%, TCI America™

Catalog No. p-7117196
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A102525G 25 g
A1025250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A102525G Supplier TCI America Supplier No. A102525G
Only null left

Chemical Identifiers

CAS 92-91-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008749
InChI Key QCZZSANNLWPGEA-UHFFFAOYSA-N
Synonym 4-acetylbiphenyl, 4'-phenylacetophenone, 4-phenylacetophenone, 4-biphenylyl methyl ketone, p-phenylacetophenone, p-acetylbiphenyl, 1-1,1'-biphenyl-4-yl ethanone, acetophenone, 4'-phenyl, 1-4-phenylphenyl ethanone, ketone, 4-biphenylyl methyl
PubChem CID 7113
IUPAC Name 1-(4-phenylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 122°C
Color White-Yellow
Boiling Point 168°C
Quantity 25 g
Formula Weight 196.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetylbiphenyl
RTECSNumber AM9662502
TSCA Yes