missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzenesulfonamide, 97%

Catalog No. AAH3290106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3290106 5 g
AAH3290103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3290106 Supplier Thermo Scientific Chemicals Supplier No. H3290106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1565-17-9 | C8H9NO3S | 199.22 g/mol

4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

Solubility
soluble in water( 2650 mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1565-17-9
Molecular Formula C8H9NO3S
Molecular Weight (g/mol) 199.22
MDL Number MFCD00792524
InChI Key CSATVXJBGFVJES-UHFFFAOYSA-N
Synonym 4-acetylbenzenesulphonamide, p-acetylbenzenesulfonamide, 4-acetyl-benzenesulfonamide, 4-acetylbenzene-1-sulfonamide, benzenesulfonamide, 4-acetyl, p-acetobenzenesulfonamide, benzenesulfonamide, p-acetyl-6ci,7ci,8ci, 4-acetyl-benzenesulfonamid, p-acetylbenzene sulfonamide, #
PubChem CID 74065
IUPAC Name 4-acetylbenzenesulfonamide
SMILES CC(=O)C1=CC=C(C=C1)S(N)(=O)=O

Specifications

Melting Point 177°C to 179°C
Quantity 5 g
Solubility Information soluble in water( 2650mg/L).
Formula Weight 199.23
Percent Purity 97%
Chemical Name or Material 4-Acetylbenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-363-5
RTECSNumber DA9386000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only