missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-N-methylaniline, 97%

Catalog No. p-7052686
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55301MD 250 mg
AAH5530103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH55301MD Supplier Thermo Scientific Chemicals Supplier No. H55301MD

CAS: 17687-47-7 | C9H11NO | 149.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17687-47-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD09261160
InChI Key GDBAYICEBDZUNM-UHFFFAOYSA-N
Synonym 4-acetyl-n-methylaniline, 1-4-methylamino phenyl ethanone, 4-acetyl-n-methyl aniline, 1-4-methylamino phenyl ethan-1-one, 1-4-methylamino-phenyl-ethanone, ethanone, 1-4-methylamino phenyl, n-methyl-4-acetylaniline, acmc-1brn7, 4-n-methylaminoacetophenone
PubChem CID 11607936
IUPAC Name 1-[4-(methylamino)phenyl]ethanone
SMILES CNC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 103°C to 107°C
Color Brown to White
Quantity 250 mg
Formula Weight 149.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Acetyl-N-methylaniline
Hazard Category H334
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P285-P304+P341-P342+P311-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only