missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-4-phenylpiperidine Hydrochloride 98.0+%, TCI America™

Catalog No. A15705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15705G Supplier TCI America Supplier No. A15705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10315-03-4
Molecular Formula C13H18ClNO
Molecular Weight (g/mol) 239.743
MDL Number MFCD00039037
InChI Key JYDHZOIDIWUHDB-UHFFFAOYSA-N
Synonym 4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride
PubChem CID 2723767
IUPAC Name 1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride
SMILES CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl

Specifications

Melting Point 248°C
Quantity 5 g
Formula Weight 239.74
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetyl-4-phenylpiperidine Hydrochloride