missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Chemical Identifiers

CAS 1807685-51-3
Molecular Formula C14H18BFO3
Molecular Weight (g/mol) 264.103
InChI Key QRPSPYZFNHVURF-UHFFFAOYSA-N
Synonym 4-Acetyl-2-fluorophenylboronic acid pinacol ester
PubChem CID 75487161
IUPAC Name 1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)C)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH6225603
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1807685-51-3
264.103
4-Acetyl-2-fluorophenylboronic acid pinacol ester
1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
C14H18BFO3
QRPSPYZFNHVURF-UHFFFAOYSA-N
75487161
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)C)F

Specifications

1807685-51-3
4-Acetyl-2-fluorophenylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)C)F
264.103
264.1
4-Acetyl-2-fluorobenzeneboronic acid pinacol ester
C14H18BFO3
QRPSPYZFNHVURF-UHFFFAOYSA-N
1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
75487161
96%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only