missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-2-chloropyridine, 97%

Catalog No. AAH3361906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3361906 5 g
AAH3361903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3361906 Supplier Thermo Scientific Chemicals Supplier No. H3361906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23794-15-2 | C7H6ClNO | 155.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23794-15-2
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.581
MDL Number MFCD07699406
InChI Key ZJCGPQZERFBGSM-UHFFFAOYSA-N
Synonym 1-2-chloropyridin-4-yl ethanone, 2-chloro-4-acetylpyridine, 4-acetyl-2-chloropyridine, 1-2-chloropyridine-4-yl ethanone, 1-2-chloropyridin-4-yl ethan-1-one, 1-2-chloro-pyridin-4-yl-ethanone, 1-2-chloro-4-pyridinyl-1-ethanone, ethanone, 1-2-chloro-4-pyridinyl, 1-2-chloro-4-pyridyl ethanone, 1-2-chloro-4-pyridinyl ethanone
PubChem CID 13145255
IUPAC Name 1-(2-chloropyridin-4-yl)ethanone
SMILES CC(=O)C1=CC(=NC=C1)Cl

Specifications

Melting Point 37°C to 40°C
Quantity 5 g
UN Number UN2811
Formula Weight 155.58
Percent Purity 97%
Chemical Name or Material 4-Acetyl-2-chloropyridine
Hazard Category H301-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only