missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetoxyacetophenone, 99%

Catalog No. AAB2228422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2228422 100 g
AAB2228414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2228422 Supplier Thermo Scientific Chemicals Supplier No. B2228422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13031-43-1 | C10H10O3 | 178.19 g/mol

4'-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.

Solubility
Slightly soluble in water(4450 mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 13031-43-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00017229
InChI Key SMIOEQSLJNNKQF-UHFFFAOYSA-N
Synonym 4'-acetoxyacetophenone, p-acetoxyacetophenone, 4-acetoxyacetophenone, p-acetylphenyl acetate, 4-acetylphenyl acetate, unii-eo1xk390jf, ethanone, 1-4-acetyloxy phenyl, 4-acetoxy acetophenone, 4-acetoxy-acetophenone, eo1xk390jf
PubChem CID 83063
ChEBI CHEBI:86558
IUPAC Name (4-acetylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 51°C to 54°C
Boiling Point 165°C to 170°C (14 mmHg)
Quantity 100 g
Beilstein 2093080
Solubility Information Slightly soluble in water(4450mg/L).
Formula Weight 178.19
Percent Purity 99%
Chemical Name or Material 4'-Acetoxyacetophenone
EINECSNumber 235-894-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only