missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxy-3-methoxycinnamic acid, 98+%

Catalog No. AAA1036103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1036103 1 g
AAA1036106 5 g
AAA1036114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1036103 Supplier Thermo Scientific Chemicals Supplier No. A1036103

CAS: 2596-47-6 | C12H12O5 | 236.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2596-47-6
Molecular Formula C12H12O5
Molecular Weight (g/mol) 236.223
MDL Number MFCD00014378
InChI Key IHKNVZISLLDMOR-GQCTYLIASA-N
Synonym acetylferulic acid, 4-acetoxy-3-methoxycinnamic acid, 3-methoxy-4-acetoxycinnamic acid, 3-4-acetoxy-3-methoxyphenyl acrylic acid, cinnamic acid, 4-acetoxy-3-methoxy, acetylated ferulic acid, cinnamic acid, 4-hydroxy-3-methoxy-, acetate, 2-propenoic acid, 3-4-acetyloxy-3-methoxyphenyl, 2e-3-4-acetyloxy-3-methoxyphenyl prop-2-enoic acid
PubChem CID 5354677
ChEBI CHEBI:86582
IUPAC Name (E)-3-(4-acetyloxy-3-methoxyphenyl)prop-2-enoic acid
SMILES CC(=O)OC1=C(C=C(C=C1)C=CC(=O)O)OC

Specifications

Melting Point 193°C to 196°C
Odor Odorless
Quantity 1 g
Beilstein 2218026
Formula Weight 236.22
Percent Purity ≥98%
Chemical Name or Material 4-Acetoxy-3-methoxycinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only