missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetamidopyridine 98.0+%, TCI America™
$150.98 - $378.40
Chemical Identifiers
| CAS | 5221-42-1 |
|---|---|
| Molecular Formula | C7H8N2O |
| Molecular Weight (g/mol) | 136.154 |
| InChI Key | BIJAWQUBRNHZGE-UHFFFAOYSA-N |
| Synonym | N-(4-Pyridyl)acetamide |
| PubChem CID | 21288 |
| IUPAC Name | N-pyridin-4-ylacetamide |
| SMILES | CC(=O)NC1=CC=NC=C1 |
Chemical Identifiers
| 5221-42-1 | |
| 136.154 | |
| N-(4-Pyridyl)acetamide | |
| N-pyridin-4-ylacetamide |
| C7H8N2O | |
| BIJAWQUBRNHZGE-UHFFFAOYSA-N | |
| 21288 | |
| CC(=O)NC1=CC=NC=C1 |
Specifications
| 5221-42-1 | |
| White-Yellow | |
| 5 g | |
| BIJAWQUBRNHZGE-UHFFFAOYSA-N | |
| N-pyridin-4-ylacetamide | |
| 21288 | |
| ≥98.0% (GC,T) | |
| 4-Acetamidopyridine |
| 152°C | |
| C7H8N2O | |
| N-(4-Pyridyl)acetamide | |
| CC(=O)NC1=CC=NC=C1 | |
| 136.154 | |
| 136.15 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : AC7875000
TSCA : No