missing translation for 'onlineSavingsMsg'
Learn More

4-(Acetamidomethyl)benzeneboronic acid, 97%

Catalog No. p-7034902
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52602MD 250 mg
AAH5260203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52602MD Supplier Thermo Scientific Chemicals Supplier No. H52602MD
Only null left

CAS: 850568-41-1 | C9H12BNO3 | 193.009 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 850568-41-1
Molecular Formula C9H12BNO3
Molecular Weight (g/mol) 193.009
MDL Number MFCD06659818
InChI Key ZMJVNKSOLIUBKO-UHFFFAOYSA-N
Synonym 4-acetamidomethyl phenyl boronic acid, 4-acetamidomethylphenylboronic acid, 4-acetamidomethylphenyl boronic acid, 4-acetamidomethyl benzeneboronic acid, 4-acetylaminomethyl benzeneboronic acid, 4-acetamidomethyl phenylboronic acid, boronic acid,b-4-acetylamino methyl phenyl, 4-acetamidomethylbenzeneboronic acid, pubchem14580
PubChem CID 22309454
IUPAC Name [4-(acetamidomethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CNC(=O)C)(O)O

Specifications

Melting Point 278°C to 280°C
Quantity 250 mg
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 4-(Acetamidomethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only