missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetamidocyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America A161725G
Specifications
| 4-Acetamidocyclohexanol (cis- and trans- mixture) | |
| 162°C | |
| C8H15NO2 | |
| 25 g | |
| CC(=O)NC1CCC(O)CC1 | |
| 157.21 | |
| 157.21 | |
| Crystalline Powder |
| 23363-88-4 | |
| White | |
| MFCD06797038,MFCD00051422 | |
| HWAFCRWGGRVEQL-UHFFFAOYSA-N | |
| N-(4-hydroxycyclohexyl)acetamide | |
| 90074 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 23363-88-4 | |
| 157.21 | |
| HWAFCRWGGRVEQL-UHFFFAOYSA-N | |
| N-(4-hydroxycyclohexyl)acetamide |
| C8H15NO2 | |
| MFCD06797038,MFCD00051422 | |
| 90074 | |
| CC(=O)NC1CCC(O)CC1 |
Safety and Handling
TSCA : No