missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidocinnamic acid, predominantly trans, 98%

Catalog No. p-7044212
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0529204 2 g
AAL0529209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0529204 Supplier Thermo Scientific Chemicals Supplier No. L0529204
Only null left

CAS: 1918352 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1918352
MDL Number MFCD00016846

Specifications

Melting Point ∼259°C (decomposition)
Molecular Formula C11H11NO3
Quantity 2 g
Beilstein 2724869
Synonym 3-4-acetylamino phenyl acrylic acid, 4-acetylaminocinnamic acid, 4-acetamidocinnamic acid, 2e-3-4-acetamidophenyl prop-2-enoic acid, 3-4-acetamidophenyl acrylic acid, 2e-3-4-acetylamino phenyl-2-propenoic acid, e-3-4-acetamidophenyl acrylic acid, 2e-3-4-acetylamino phenyl acrylic acid, e-3-4-acetamidophenyl prop-2-enoic acid, 2e-3-4-acetylamino phenyl prop-2-enoic acid
InChI Key WGMFHSADKZJPGR-QPJJXVBHSA-N
SMILES CC(=O)NC1=CC=C(C=C1)C=CC(=O)O
IUPAC Name (E)-3-(4-acetamidophenyl)prop-2-enoic acid
Molecular Weight (g/mol) 205.21
PubChem CID 5357155
Formula Weight 205.21
Percent Purity 0.98
Chemical Name or Material 4-Acetamidocinnamic acid, predominantly trans
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only