missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetamidoacetophenone 98.0+%, TCI America™

Catalog No. A28215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28215G 5 g
A282125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28215G Supplier TCI America Supplier No. A28215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2719-21-3
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD00014965
InChI Key WECHHDJTILFYQT-UHFFFAOYSA-N
Synonym 4'-acetamidoacetophenone, n-4-acetylphenyl acetamide, 4-acetamidoacetophenone, n-p-acetylphenyl acetamide, acetamide, n-4-acetylphenyl, p-acetylaminoacetophenone, 4-acetylacetanilide, 4'-acetylacetanilide, p-acetamidoacetophenone, 4-acetamido-acetophenone
PubChem CID 75937
IUPAC Name N-(4-acetylphenyl)acetamide
SMILES CC(=O)C1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 171°C
Quantity 5 g
Formula Weight 177.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Acetamidoacetophenone