missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidoacetophenone, 98%, Thermo Scientific™

Chemical Identifiers

CAS 2719-21-3
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00014965
InChI Key WECHHDJTILFYQT-UHFFFAOYSA-N
Synonym 4'-acetamidoacetophenone, n-4-acetylphenyl acetamide, 4-acetamidoacetophenone, n-p-acetylphenyl acetamide, acetamide, n-4-acetylphenyl, p-acetylaminoacetophenone, 4-acetylacetanilide, 4'-acetylacetanilide, p-acetamidoacetophenone, 4-acetamido-acetophenone
PubChem CID 75937
IUPAC Name N-(4-acetylphenyl)acetamide
SMILES CC(=O)C1=CC=C(C=C1)NC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391820050
SDS View Documents
Thermo Scientific Chemicals
391820050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2719-21-3
177.2
WECHHDJTILFYQT-UHFFFAOYSA-N
75937
CC(=O)C1=CC=C(C=C1)NC(=O)C
C10H11NO2
MFCD00014965
4'-acetamidoacetophenone, n-4-acetylphenyl acetamide, 4-acetamidoacetophenone, n-p-acetylphenyl acetamide, acetamide, n-4-acetylphenyl, p-acetylaminoacetophenone, 4-acetylacetanilide, 4'-acetylacetanilide, p-acetamidoacetophenone, 4-acetamido-acetophenone
N-(4-acetylphenyl)acetamide

Specifications

2719-21-3
100.0
97.5% min. (HPLC)
C10H11NO2
MFCD00014965
4'-acetamidoacetophenone, n-4-acetylphenyl acetamide, 4-acetamidoacetophenone, n-p-acetylphenyl acetamide, acetamide, n-4-acetylphenyl, p-acetylaminoacetophenone, 4-acetylacetanilide, 4'-acetylacetanilide, p-acetamidoacetophenone, 4-acetamido-acetophenone
WECHHDJTILFYQT-UHFFFAOYSA-N
N-(4-acetylphenyl)acetamide
75937
98%
97.5
Authentic
Glass bottle
CH3COC6H4NHCOCH3
5g
Solubility in water: almost insoluble. Other solubilities: soluble in benzene
CC(=O)C1=CC=C(C=C1)NC(=O)C
177.2
177.2
4-Acetamidoacetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 220-320-6

SDS
Documents

Documents

RUO – Research Use Only