missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-TEMPO, free radical, 98+%

Catalog No. AAB2345606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2345606 5 g
AAB2345603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2345606 Supplier Thermo Scientific Chemicals Supplier No. B2345606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14691-89-5 | C11H21N2O2 | 213.301 g/mol

4-Acetamido-TEMPO, free radical oxidizes alcohols to carbonyl compounds in the presence of TsOH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetamido-TEMPO, free radical oxidizes alcohols to carbonyl compounds in the presence of TsOH.

Solubility
Soluble in ethanol, acetone, acetonitrile, methylene chloride. Insoluble in hexane, ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, heat.

Chemical Identifiers

CAS 14691-89-5
Molecular Formula C11H21N2O2
Molecular Weight (g/mol) 213.301
MDL Number MFCD00043593
InChI Key UXBLSWOMIHTQPH-UHFFFAOYSA-N
Synonym 4-acetamido-tempo, 4-acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-acetamido-tempo, free radical, unii-g7abj73l6s, g7abj73l6s, 4-acetylamino-2,2,6,6-tetramethylpiperidin-1-oxyl, pubchem9193, aa-tempo, 4-acetamido-2,2,6,6-tetramethyl-1-piperidinyl oxidanyl
PubChem CID 518988
IUPAC Name N-(1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl)acetamide
SMILES CC(=O)NC1CC(N(C(C1)(C)C)[O])(C)C

Specifications

Melting Point 143°C to 146°C
Quantity 5 g
Beilstein 3546225
Solubility Information Soluble in ethanol,acetone,acetonitrile,methylene chloride. Insoluble in hexane,ether.
Formula Weight 213.3
Percent Purity ≥98%
Chemical Name or Material 4-Acetamido-TEMPO, free radical
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 423-840-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only