missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetamido-N-(2-aminophenyl)benzamide 98.0+%, TCI America™
Supplier: TCI America A250110MG
Specifications
| 4-Acetamido-N-(2-aminophenyl)benzamide | |
| 242°C | |
| C15H15N3O2 | |
| 10 mg | |
| VAZAPHZUAVEOMC-UHFFFAOYSA-N | |
| N-(2-aminophenyl)-4-acetamidobenzamide | |
| 2746 | |
| ≥98.0% (HPLC) |
| 112522-64-2 | |
| Yellow | |
| MFCD00866266 | |
| tacedinaline, acetyldinaline, 4-acetamido-n-2-aminophenyl benzamide, 4-acetylamino-n-2'-aminophenyl benzamide, tacedinalina, ci994 tacedinaline, n-acetyldinaline, tacedinaline usan:inn, tacedinalina inn-spanish, 4-acetylamino-n-2-aminophenyl benzamide | |
| CC(=O)NC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1N | |
| 269.30 | |
| 269.30 | |
| Crystalline Powder |
Chemical Identifiers
| 112522-64-2 | |
| 269.30 | |
| VAZAPHZUAVEOMC-UHFFFAOYSA-N | |
| 2746 | |
| CC(=O)NC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1N |
| C15H15N3O2 | |
| MFCD00866266 | |
| tacedinaline, acetyldinaline, 4-acetamido-n-2-aminophenyl benzamide, 4-acetylamino-n-2'-aminophenyl benzamide, tacedinalina, ci994 tacedinaline, n-acetyldinaline, tacedinaline usan:inn, tacedinalina inn-spanish, 4-acetylamino-n-2-aminophenyl benzamide | |
| N-(2-aminophenyl)-4-acetamidobenzamide |
Safety and Handling
RTECSNumber : CU8702023
TSCA : No
Recommended Storage : Refrigerator