missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-N-(2-aminophenyl)benzamide 98.0+%, TCI America™

Catalog No. A250150MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A250150MG 50 mg
A250110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A250150MG Supplier TCI America Supplier No. A250150MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112522-64-2
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
MDL Number MFCD00866266
InChI Key VAZAPHZUAVEOMC-UHFFFAOYSA-N
Synonym tacedinaline, acetyldinaline, 4-acetamido-n-2-aminophenyl benzamide, 4-acetylamino-n-2'-aminophenyl benzamide, tacedinalina, ci994 tacedinaline, n-acetyldinaline, tacedinaline usan:inn, tacedinalina inn-spanish, 4-acetylamino-n-2-aminophenyl benzamide
PubChem CID 2746
IUPAC Name N-(2-aminophenyl)-4-acetamidobenzamide
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1N

Specifications

Melting Point 242°C
Color Yellow
Quantity 50 mg
Formula Weight 269.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-N-(2-aminophenyl)benzamide
RTECSNumber CU8702023
TSCA No
Recommended Storage Refrigerator