missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 4-Acetamido-4'-Maleimidylstilbene-2,2'-Disulfonic Acid, Disodium Salt

Catalog No. a485
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A485 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A485 Supplier Invitrogen™ Supplier No. A485
Only null left

4-acetamido-4'-maleimidylstilbene-2,2'-disulfonic acid, disodium salt

Our 4-acetamido-4'-maleimidylstilbene-2,2'- disulfonic acid is a thiol-reactive reagent that is water soluble, with high polarity and membrane impermeability. This polysulfonated dye is useful for determining whether thiol-containing proteins and polypeptide chains are exposed at the extracellular or cytoplasmic membrane surface.

Specifications

Quantity 25 mg
Product Type 4-Acetamido-4'-Maleimidylstilbene-2,2'-Disulfonic Acid
Content And Storage Store in freezer (-5 to -30°C).
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye Stilbene
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.