missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetamido-3'-bromoacetophenone 98.0+%, TCI America™

Catalog No. A28525G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A28525G Supplier TCI America Supplier No. A28525G
Only null left

Chemical Identifiers

CAS 101209-08-9
Molecular Formula C10H10BrNO2
Molecular Weight (g/mol) 256.10
MDL Number MFCD00051781
InChI Key PMYJAVHDFDKJBS-UHFFFAOYSA-N
Synonym 4-acetamido-3-bromoacetophenone, 4'-acetamido-3'-bromoacetophenone, n-4-acetyl-2-bromophenyl acetamide, 4-acetyl-2-bromoacetanilide, 4'-acetamido-3-bromoacetophenone, 3'-bromo-4'-acetaminoacetophenone, n-2-bromo-4-acetylphenyl acetamide, n-4-acetyl-2-bromo-phenyl acetamide, n-4-acetyl-2-bromo-phenyl-acetamide
PubChem CID 3682474
IUPAC Name N-(4-acetyl-2-bromophenyl)acetamide
SMILES CC(=O)NC1=C(Br)C=C(C=C1)C(C)=O

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 5 g
Formula Weight 256.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Acetamido-3′-bromoacetophenone