missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™

Catalog No. p-7117383
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13485G 5 g
A134825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13485G Supplier TCI America Supplier No. A13485G

Chemical Identifiers

CAS 14691-89-5
Molecular Formula C11H21N2O2
Molecular Weight (g/mol) 213.301
MDL Number MFCD00043593
InChI Key UXBLSWOMIHTQPH-UHFFFAOYSA-N
Synonym 4-acetamido-tempo, 4-acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-acetamido-tempo, free radical, unii-g7abj73l6s, g7abj73l6s, 4-acetylamino-2,2,6,6-tetramethylpiperidin-1-oxyl, pubchem9193, aa-tempo, 4-acetamido-2,2,6,6-tetramethyl-1-piperidinyl oxidanyl
PubChem CID 518988
IUPAC Name N-(1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl)acetamide
SMILES CC(=O)NC1CC(N(C(C1)(C)C)[O])(C)C

Specifications

Melting Point 143°C
Quantity 5 g
Formula Weight 213.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical [Catalyst for Oxidation]