missing translation for 'onlineSavingsMsg'
Learn More

4,8-Bis(dodecyloxy)benzo[1,2-b:4,5-b']dithiophene 98.0+%, TCI America™

Catalog No. b54541g
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54541G 1 g
B5454200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54541G Supplier TCI America Supplier No. B54541G
Only null left

Chemical Identifiers

CAS 1044795-04-1
Molecular Formula C34H54O2S2

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 558.92
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,8-Bis(dodecyloxy)benzo[1,2-b:4,5-b′]dithiophene