missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dimethyl-1,10-phenanthroline, 98%

Catalog No. AAA1777901
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1777901 0.25 g
AAA1777903 1 g
AAA1777906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1777901 Supplier Thermo Scientific Chemicals Supplier No. A1777901
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3248-05-3 | C14H12N2 | 208.26 g/mol

4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.

Solubility
Soluble in benzene and alcohol. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and acids.

Chemical Identifiers

CAS 3248-05-3
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.26
MDL Number MFCD00004979
InChI Key JIVLDFFWTQYGSR-UHFFFAOYSA-N
Synonym 4,7-dimethyl-o-phenanthroline, 1,10-phenanthroline, 4,7-dimethyl, 4,7-dimethyl-1,10 phenanthroline, 4,7-dimethylpyridino 3,2-h quinoline, acmc-1cjn6, bis 5-chlorophenyl acetic acid, 4,7-dimethyl-1,9-phenanthroline, 1,10-phenanthroline, 4,7-dimethyl-, hydrate
PubChem CID 72792
IUPAC Name 4,7-dimethyl-1,10-phenanthroline
SMILES CC1=C2C=CC3=C(C=CN=C3C2=NC=C1)C

Specifications

Melting Point 193°C to 195°C
Quantity 0.25 g
Solubility Information Soluble in benzene and alcohol. Slightly soluble in water.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material 4,7-Dimethyl-1,10-phenanthroline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 221-827-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only