missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dihydroxy-1,10-phenanthroline

Catalog No. p-7049931
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3143402 0.5 g
AA3143404 2 g
AA3143409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3143402 Supplier Thermo Scientific Chemicals Supplier No. 03143402

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3922-40-5 | C12H8N2O2 | 212.208 g/mol

4,7-Dihydroxy-1,10-phenanthroline is used as a phenanthroline stain. Its iron(II) complex reacts rapidly with dissolved oxygen in aqueous solution, thereby it is utilized for the spectrophotometric determination of dissolved oxygen up to 20 ppm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3922-40-5
Molecular Formula C12H8N2O2
Molecular Weight (g/mol) 212.208
MDL Number MFCD00004975
InChI Key SLIBCJURSADKPV-UHFFFAOYSA-N
Synonym 1,10-phenanthroline-4,7-diol, 4,7-dihydroxy-1,10-phenanthroline, 1,10-phenanthroline-4,7 1h,10h-dione, 1,4,7,10-tetrahydro-1,10-phenanthroline-4,7-dione, pyridino 3,2-h quinoline-4,7-diol, acmc-1coeh, bidd:gt0532, 1,1-phenanthroline-4,7-diol, 1,10 phenanthroline-4,7-diol, 4,7-dioxylato-1,10-phenanthroline
PubChem CID 77524
IUPAC Name 1,10-dihydro-1,10-phenanthroline-4,7-dione
SMILES C1=CC2=C(C3=C1C(=O)C=CN3)NC=CC2=O

Specifications

Melting Point >300°C (decomposition)
Color Yellow
Quantity 0.5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 212.21
Physical Form Powder
Chemical Name or Material 4,7-Dihydroxy-1,10-phenanthroline
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 223-493-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only