missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dichloroisatin 97.0+%, TCI America™

Catalog No. D40395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40395G 5 g
D40391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40395G Supplier TCI America Supplier No. D40395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18711-13-2
Molecular Formula C8H3Cl2NO2
Molecular Weight (g/mol) 216.017
MDL Number MFCD00047214
InChI Key NUXYYWOWNFEMNH-UHFFFAOYSA-N
Synonym 4,7-dichloroisatin, 4,7-dichloroindoline-2,3-dione, 4,7-dichloro isatin, 4,7-dichloro-isatin, 4,7-dichloro-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 4,7-dichloro, 4,7-dichloroisatin;, 4,7-dichloroisatine;, d0ek7h, isatin-based compound, 51
PubChem CID 2734793
IUPAC Name 4,7-dichloro-1H-indole-2,3-dione
SMILES C1=CC(=C2C(=C1Cl)C(=O)C(=O)N2)Cl

Specifications

Melting Point 252°C
Quantity 5 g
Formula Weight 216.02
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dichloroisatin