missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dichloroisatin, 98%

Catalog No. AAA1485306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1485306 5 g
AAA1485303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1485306 Supplier Thermo Scientific Chemicals Supplier No. A1485306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18711-13-2 | C8H3Cl2NO2 | 216.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18711-13-2
Molecular Formula C8H3Cl2NO2
Molecular Weight (g/mol) 216.017
MDL Number MFCD00047214
InChI Key NUXYYWOWNFEMNH-UHFFFAOYSA-N
Synonym 4,7-dichloroisatin, 4,7-dichloroindoline-2,3-dione, 4,7-dichloro isatin, 4,7-dichloro-isatin, 4,7-dichloro-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 4,7-dichloro, 4,7-dichloroisatin;, 4,7-dichloroisatine;, d0ek7h, isatin-based compound, 51
PubChem CID 2734793
IUPAC Name 4,7-dichloro-1H-indole-2,3-dione
SMILES C1=CC(=C2C(=C1Cl)C(=O)C(=O)N2)Cl

Specifications

Melting Point 246°C to 249°C
Quantity 5 g
Beilstein 168731
Formula Weight 216.02
Percent Purity 98%
Chemical Name or Material 4,7-Dichloroisatin
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only