missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine 97.0+%, TCI America™
$145.40 - $434.32
Chemical Identifiers
| CAS | 333432-27-2 |
|---|---|
| Molecular Formula | C5HBr2N3S |
| Molecular Weight (g/mol) | 294.952 |
| MDL Number | MFCD23703147 |
| InChI Key | LEHZIBSAFRVAJP-UHFFFAOYSA-N |
| PubChem CID | 58129789 |
| IUPAC Name | 4,7-dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine |
| SMILES | C1=C(C2=NSN=C2C(=N1)Br)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4442200MG
|
TCI America
D4442200MG |
200 mg |
Each for $145.40
|
|
|||||
|
D44421G
|
TCI America
D44421G |
1 g |
Each for $434.32
|
|
|||||
Chemical Identifiers
| 333432-27-2 | |
| 294.952 | |
| LEHZIBSAFRVAJP-UHFFFAOYSA-N | |
| 4,7-dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine |
| C5HBr2N3S | |
| MFCD23703147 | |
| 58129789 | |
| C1=C(C2=NSN=C2C(=N1)Br)Br |
Specifications
| 333432-27-2 | |
| Yellow | |
| MFCD23703147 | |
| LEHZIBSAFRVAJP-UHFFFAOYSA-N | |
| 4,7-dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine | |
| 58129789 | |
| ≥97.0% (GC) | |
| 4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine |
| 148°C | |
| C5HBr2N3S | |
| 200 mg | |
| C1=C(C2=NSN=C2C(=N1)Br)Br | |
| 294.952 | |
| 294.95 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator