missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dibromo-1,10-phenanthroline Hydrate 98.0+%, TCI America™

Catalog No. p-7128588
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4987200MG 200 mg
D49871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4987200MG Supplier TCI America Supplier No. D4987200MG
Only null left

Chemical Identifiers

CAS 156492-30-7
Molecular Formula C12H6Br2N2
Molecular Weight (g/mol) 338.002
MDL Number MFCD25562940
InChI Key AKZAIDYHEKUXBU-UHFFFAOYSA-N
PubChem CID 11393583
IUPAC Name 4,7-dibromo-1,10-phenanthroline
SMILES C1=CC2=C(C=CN=C2C3=NC=CC(=C31)Br)Br

Specifications

Melting Point 236°C
Color Yellow
Quantity 200 mg
Formula Weight 338.00
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-1,10-phenanthroline Hydrate