missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole 95.0+%, TCI America™
$111.84 - $469.75
Chemical Identifiers
| CAS | 288071-87-4 |
|---|---|
| Molecular Formula | C14H6Br2N2S3 |
| Molecular Weight (g/mol) | 458.204 |
| MDL Number | MFCD16619295 |
| InChI Key | ZIIMIGRZSUYQGW-UHFFFAOYSA-N |
| PubChem CID | 22381654 |
| IUPAC Name | 4,7-bis(5-bromothiophen-2-yl)-2,1,3-benzothiadiazole |
| SMILES | C1=C(C2=NSN=C2C(=C1)C3=CC=C(S3)Br)C4=CC=C(S4)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3886100MG
|
TCI America
B3886100MG |
100 mg |
Each for $111.84
|
|
|||||
|
B38861G
|
TCI America
B38861G |
1 g |
Each for $469.75
|
|
|||||
Chemical Identifiers
| 288071-87-4 | |
| 458.204 | |
| ZIIMIGRZSUYQGW-UHFFFAOYSA-N | |
| 4,7-bis(5-bromothiophen-2-yl)-2,1,3-benzothiadiazole |
| C14H6Br2N2S3 | |
| MFCD16619295 | |
| 22381654 | |
| C1=C(C2=NSN=C2C(=C1)C3=CC=C(S3)Br)C4=CC=C(S4)Br |
Specifications
| 288071-87-4 | |
| C14H6Br2N2S3 | |
| 100 mg | |
| C1=C(C2=NSN=C2C(=C1)C3=CC=C(S3)Br)C4=CC=C(S4)Br | |
| 458.204 | |
| 458.20 | |
| Crystalline Powder |
| 247°C | |
| MFCD16619295 | |
| ZIIMIGRZSUYQGW-UHFFFAOYSA-N | |
| 4,7-bis(5-bromothiophen-2-yl)-2,1,3-benzothiadiazole | |
| 22381654 | |
| ≥95.0% (HPLC,N) | |
| 4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole |
Safety and Handling
TSCA : No