missing translation for 'onlineSavingsMsg'
Learn More

4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole 95.0+%, TCI America™

Catalog No. B38861G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38861G 1 g
B3886100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38861G Supplier TCI America Supplier No. B38861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 288071-87-4
Molecular Formula C14H6Br2N2S3
Molecular Weight (g/mol) 458.204
MDL Number MFCD16619295
InChI Key ZIIMIGRZSUYQGW-UHFFFAOYSA-N
PubChem CID 22381654
IUPAC Name 4,7-bis(5-bromothiophen-2-yl)-2,1,3-benzothiadiazole
SMILES C1=C(C2=NSN=C2C(=C1)C3=CC=C(S3)Br)C4=CC=C(S4)Br

Specifications

Melting Point 247°C
Quantity 1 g
Formula Weight 458.20
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole