missing translation for 'onlineSavingsMsg'
Learn More

4-[6-(Tributylstannyl)-2-pyridinyl]morpholine, 95%, Thermo Scientific™

Catalog No. CC38341CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC38341CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC38341CB Supplier Thermo Scientific Chemicals Supplier No. CC38341CB

Chemical Identifiers

CAS 869901-24-6
Molecular Formula C21H38N2OSn
Molecular Weight (g/mol) 453.258
MDL Number MFCD08271894
InChI Key LDERBIPTTICDSG-UHFFFAOYSA-N
Synonym 4-6-tributylstannyl pyridin-2-yl morpholine, 4-6-tributylstannyl-2-pyridinyl morpholine, pubchem21425, 4-6-tributylstannanyl-pyridin-2-yl-morpholine, morpholine,4-6-tributylstannyl-2-pyridinyl
PubChem CID 18525739
IUPAC Name tributyl-(6-morpholin-4-ylpyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)N2CCOCC2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 453.25
Percent Purity 95%
Chemical Name or Material 4-[6-(Tributylstannyl)-2-pyridinyl]morpholine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.