missing translation for 'onlineSavingsMsg'
Learn More

4,6-O-Isopropylidene-D-glucal, 97%, Thermo Scientific Chemicals

Catalog No. p-7040446
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60221MD 250 mg
AAH6022103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60221MD Supplier Thermo Scientific Chemicals Supplier No. H60221MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51450-36-3 | Molecular Formula: C9H14O4 | Molecular Weight: 186.207 g/mol

4,6-O-Isopropylidene-D-glucal is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-O-Isopropylidene-D-glucal is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 51450-36-3
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD22988998
InChI Key GAQDIYMHBQNXLE-PRJMDXOYSA-N
Synonym 4,6-o-isopropylidene-d-glucal, 4-o,6-o-isopropylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,2-didehydro-1,2-dideoxy-4-o,6-o-isopropylidene-beta-d-glucopyranose, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol
PubChem CID 11052343
IUPAC Name (4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
SMILES CC1(OCC2C(O1)C(C=CO2)O)C

Specifications

Quantity 250 mg
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4,6-O-Isopropylidene-D-glucal
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only