missing translation for 'onlineSavingsMsg'
Learn More

4,6-O-Ethylidene-alpha-D-glucopyranose 92.0+%, TCI America™

Catalog No. e04025g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04025G Supplier TCI America Supplier No. E04025G

Specifications

CAS 13224-99-2
Melting Point 173°C
Color White-Yellow
Molecular Formula C8H14O6
Quantity 5 g
MDL Number MFCD00006820
Synonym 4,6-o-ethylidene-a-d-glucose
InChI Key VZPBLPQAMPVTFO-UHFFFAOYNA-N
SMILES CC1OCC2OC(O)C(O)C(O)C2O1
IUPAC Name 2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxine-6,7,8-triol
Molecular Weight (g/mol) 206.19
PubChem CID 21581900
Formula Weight 206.19
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-O-Ethylidene-alpha-D-glucopyranose
Show More Show Less