missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(6-Hydroxyhexyloxy)benzoic Acid 97.0+%, TCI America™
$154.71 - $469.75
Chemical Identifiers
| CAS | 83883-25-4 |
|---|---|
| Molecular Formula | C13H18O4 |
| Molecular Weight (g/mol) | 238.283 |
| MDL Number | MFCD00971624 |
| InChI Key | VVYHWYFUTOHXRH-UHFFFAOYSA-N |
| PubChem CID | 3628413 |
| IUPAC Name | 4-(6-hydroxyhexoxy)benzoic acid |
| SMILES | C1=CC(=CC=C1C(=O)O)OCCCCCCO |
Chemical Identifiers
| 83883-25-4 | |
| 238.283 | |
| VVYHWYFUTOHXRH-UHFFFAOYSA-N | |
| 4-(6-hydroxyhexoxy)benzoic acid |
| C13H18O4 | |
| MFCD00971624 | |
| 3628413 | |
| C1=CC(=CC=C1C(=O)O)OCCCCCCO |
Specifications
| 83883-25-4 | |
| White | |
| MFCD00971624 | |
| VVYHWYFUTOHXRH-UHFFFAOYSA-N | |
| 4-(6-hydroxyhexoxy)benzoic acid | |
| 3628413 | |
| ≥97.0% (GC) | |
| 4-(6-Hydroxyhexyloxy)benzoic Acid |
| 139°C | |
| C13H18O4 | |
| 1 g | |
| C1=CC(=CC=C1C(=O)O)OCCCCCCO | |
| 238.283 | |
| 238.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No