missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Hydroxyhexyloxy)benzoic Acid 97.0+%, TCI America™

Catalog No. H16295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16295G 5 g
H16291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16295G Supplier TCI America Supplier No. H16295G
Only null left

Chemical Identifiers

CAS 83883-25-4
Molecular Formula C13H18O4
Molecular Weight (g/mol) 238.283
MDL Number MFCD00971624
InChI Key VVYHWYFUTOHXRH-UHFFFAOYSA-N
PubChem CID 3628413
IUPAC Name 4-(6-hydroxyhexoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OCCCCCCO

Specifications

Melting Point 139°C
Color White
Quantity 5 g
Formula Weight 238.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(6-Hydroxyhexyloxy)benzoic Acid