missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diphenyl-2-pyrone, 98%

Catalog No. p-7081682
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1175503 1 g
AAA1175506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1175503 Supplier Thermo Scientific Chemicals Supplier No. A1175503

CAS: 17372-52-0 | C17H12O2 | 248.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17372-52-0
Molecular Formula C17H12O2
Molecular Weight (g/mol) 248.28
MDL Number MFCD00031018
InChI Key DDGIHXFDWLCKRZ-UHFFFAOYSA-N
PubChem CID 296685
IUPAC Name 4,6-diphenylpyran-2-one
SMILES O=C1OC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C to 138°C
Odor Odorless
Quantity 1 g
Synonym 4,6-diphenyl-2h-pyran-2-one, 4,6-diphenyl-2-pyrone, 2h-pyran-2-one,4,6-diphenyl, pubchem13218, maybridge1_002884, 4,6-diphenyl-2-pyranone, 2h-pyran-2-one, 4,6-diphenyl
Formula Weight 248.27
Percent Purity 98%
Chemical Name or Material 4,6-Diphenyl-2-pyrone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only