missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxypyrimidine, 98%

CAS: 1193-24-4 | C4H4N2O2 | 112.09 g/mol

$253.03 - $253.03

Chemical Identifiers

CAS 1193-24-4
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00016733
InChI Key DUFGYCAXVIUXIP-UHFFFAOYSA-N
Synonym 4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
PubChem CID 14512
SMILES OC1=CC(=O)NC=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC115081000
SDS View Documents
Thermo Scientific Chemicals
115081000
100 g Plastic bottle
Each for $253.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1193-24-4
112.09
DUFGYCAXVIUXIP-UHFFFAOYSA-N
14512
C4H4N2O2
MFCD00016733
4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
OC1=CC(=O)NC=N1

Specifications

1193-24-4
100.0
Yellow to Orange
98%
C4H4N2O2
100 g
4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
DUFGYCAXVIUXIP-UHFFFAOYSA-N
4-hydroxy-1H-pyrimidin-6-one
14512
98%
4, 6-Dihydroxypyrimidine
97.5
>300°C
Authentic
Plastic bottle
MFCD00016733
26,444
Solubility in water: 2.5g/L water (20° c)
OC1=CC(=O)NC=N1
112.09
112.09
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-772-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only