missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine 98.0+%, TCI America™

Catalog No. p-7127329
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24885G 5 g
D248825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24885G Supplier TCI America Supplier No. D24885G
Only null left

Chemical Identifiers

CAS 1193-21-1
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00006109
InChI Key XJPZKYIHCLDXST-UHFFFAOYSA-N
Synonym pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine
PubChem CID 70943
IUPAC Name 4,6-dichloropyrimidine
SMILES ClC1=CC(Cl)=NC=N1

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
UN Number 3263
Formula Weight 148.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloropyrimidine
EINECSNumber (5)-0927
TSCA No