Learn More
4,6-Dichloropyrimidine, 97%
CAS: 1193-21-1 | C4H2Cl2N2 | 148.97 g/mol
$282.44 - $282.44
Chemical Identifiers
| CAS | 1193-21-1 |
|---|---|
| Molecular Formula | C4H2Cl2N2 |
| Molecular Weight (g/mol) | 148.97 |
| MDL Number | MFCD00006109 |
| InChI Key | XJPZKYIHCLDXST-UHFFFAOYSA-N |
| Synonym | pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine |
| PubChem CID | 70943 |
| IUPAC Name | 4,6-dichloropyrimidine |
| SMILES | ClC1=CC(Cl)=NC=N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC163370250
|
Thermo Scientific Chemicals
163370250 |
25 g | Glass bottle |
Each for $282.44
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1193-21-1 | |
| 148.97 | |
| XJPZKYIHCLDXST-UHFFFAOYSA-N | |
| 70943 | |
| ClC1=CC(Cl)=NC=N1 |
| C4H2Cl2N2 | |
| MFCD00006109 | |
| pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine | |
| 4,6-dichloropyrimidine |
Specifications
| 1193-21-1 | |
| 100.0 | |
| Yellow to Green | |
| Authentic | |
| Glass bottle | |
| MFCD00006109 | |
| pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine | |
| ClC1=CC(Cl)=NC=N1 | |
| 148.97 | |
| 148.98 | |
| Crystals |
| 96.0 | |
| 63°C to 68°C | |
| 176°C | |
| 96% min. (GC) | |
| C4H2Cl2N2 | |
| 25 g | |
| XJPZKYIHCLDXST-UHFFFAOYSA-N | |
| 4,6-dichloropyrimidine | |
| 70943 | |
| 97% | |
| 4, 6-Dichloropyrimidine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several m
GHS Signal Word: Danger
EINECSNumber : 214-770-2
RUO – Research Use Only