missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine, 97%

Catalog No. AC163370050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163370050 5 g Glass bottle
AC163370250 25 g Glass bottle
AC163371000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163370050 Supplier Thermo Scientific Chemicals Supplier No. 163370050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1193-21-1 | C4H2Cl2N2 | 148.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1193-21-1
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00006109
InChI Key XJPZKYIHCLDXST-UHFFFAOYSA-N
Synonym pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine
PubChem CID 70943
IUPAC Name 4,6-dichloropyrimidine
SMILES ClC1=CC(Cl)=NC=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63°C to 68°C
Color Yellow to Green
Boiling Point 176°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 148.98
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 4, 6-Dichloropyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several m
EINECSNumber 214-770-2

RUO – Research Use Only