missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine, 97%

CAS: 1193-21-1 | C4H2Cl2N2 | 148.97 g/mol

$270.10 - $270.10

Chemical Identifiers

CAS 1193-21-1
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00006109
InChI Key XJPZKYIHCLDXST-UHFFFAOYSA-N
Synonym pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine
PubChem CID 70943
IUPAC Name 4,6-dichloropyrimidine
SMILES ClC1=CC(Cl)=NC=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1321214
View Documents
Thermo Scientific Chemicals
L1321214
25 g
Each for $270.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1193-21-1
148.97
XJPZKYIHCLDXST-UHFFFAOYSA-N
70943
ClC1=CC(Cl)=NC=N1
C4H2Cl2N2
MFCD00006109
pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine
4,6-dichloropyrimidine

Specifications

1193-21-1
176°C
MFCD00006109
UN3261
pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine
ClC1=CC(Cl)=NC=N1
148.97
148.98
4,6-Dichloropyrimidine
64°C to 68°C
C4H2Cl2N2
25 g
111195
XJPZKYIHCLDXST-UHFFFAOYSA-N
4,6-dichloropyrimidine
70943
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H312+H332-H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 214-770-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only