missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-nitropyrimidine, 98%

Catalog No. AAH5625203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5625203 1 g
AAH5625206 5 g
AAH5625214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5625203 Supplier Thermo Scientific Chemicals Supplier No. H5625203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4316-93-2 | C4HCl2N3O2 | 193.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4316-93-2
Molecular Formula C4HCl2N3O2
Molecular Weight (g/mol) 193.97
MDL Number MFCD00006107
InChI Key HCTISZQLTGAYOX-UHFFFAOYSA-N
Synonym pyrimidine, 4,6-dichloro-5-nitro, 4,6-dichloro-5-nitro-pyrimidine, 4,6-dichloro-5-nitro-pyridine, pubchem7078, acmc-1an8c, 4-23-00-00899 beilstein handbook reference, ksc495i5h, 4,6-dichloro-5-nitroprimidine, 4,6 dichloro-5-nitropyrimidine, 5-nitro 4,6-dichloropyrimidine
PubChem CID 20312
IUPAC Name 4,6-dichloro-5-nitropyrimidine
SMILES [O-][N+](=O)C1=C(Cl)N=CN=C1Cl

Specifications

Melting Point 100°C to 103°C
Quantity 1 g
Beilstein 162029
Formula Weight 193.98
Percent Purity 98%
Chemical Name or Material 4,6-Dichloro-5-nitropyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-340-6
RTECSNumber UV8258000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only