missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine 98.0+%, TCI America™

Catalog No. D45985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45985G 5 g
D45981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45985G Supplier TCI America Supplier No. D45985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150728-13-5
Molecular Formula C15H10Cl2N4O2
Molecular Weight (g/mol) 349.171
MDL Number MFCD03839838
InChI Key IZGOBGVYADHVKH-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-2-methoxyphenoxy-2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy-2,2-bipyrimidine, 2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy, 4,6-dichloro-5-2-methoxy-phenoxy-2,2' bipyrimidinyl, 2,2'-bipyrimidine,4,6-dichloro-5-2-methoxyphenoxy, 4,6-dichloro-5-2-methoxyphenoxy-2-pyrimidin-2-yl pyrimidine, 4,6-dichloro-5-2-methoxyphenoxy 2,2'-bipyrimidine, 4,6-dichloro-5-o-methoxyphenoxy-2-2-pyrimidinyl pyrimidine, pubchem17752
PubChem CID 10545706
IUPAC Name 4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
SMILES COC1=CC=CC=C1OC2=C(N=C(N=C2Cl)C3=NC=CC=N3)Cl

Specifications

Melting Point 159°C
Color Yellow
Quantity 5 g
Formula Weight 349.17
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2′-bipyrimidine