missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dibromodibenzofuran 98.0+%, TCI America™

Catalog No. p-7128506
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4858200MG 200 mg
D48581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4858200MG Supplier TCI America Supplier No. D4858200MG

Chemical Identifiers

CAS 201138-91-2
Molecular Formula C12H6Br2O
Molecular Weight (g/mol) 325.987
MDL Number MFCD00185686
InChI Key XSJLDNSNUICSQC-UHFFFAOYSA-N
PubChem CID 526362
IUPAC Name 4,6-dibromodibenzofuran
SMILES C1=CC2=C(C(=C1)Br)OC3=C2C=CC=C3Br

Specifications

Melting Point 149°C
Color White
Quantity 200 mg
Formula Weight 325.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dibromodibenzofuran